Learn More
Brands LABELING KIT
Catalog No. NC9080267
Encompass
Convenient fluorescent labeling of glycans with 2-AB (2-aminobenzamide) by reductive amination. Any purified glycan or glycan pool with a free reducing sugar may be labeled. Up to 36 samples may be processed (2 batches of 18). Cleanup columns and cartridges must be purchased separately.
Catalog No. NC9080267 Supplier Brands Supplier No. GKK404*
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Convenient fluorescent labeling of glycans with 2-AB (2-aminobenzamide) by reductive amination. Any purified glycan or glycan pool with a free reducing sugar may be labeled. Up to 36 samples may be processed (2 batches of 18). Cleanup columns and cartridges must be purchased separately.