missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ WaterPro™ Station Reverse Osmosis Membrane

Catalog No. 16307102
Change view
Click to view available options
Feedwater Source:
Tap
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Feedwater Source
16307102 Tap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16307102 Supplier Labconco™ Supplier No. 9078700

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reverse Osmosis Membrane required for operation of WaterPro RO Water Purification Systems.

Reverse Osmosis Membrane required for operation of Labconco WaterPro RO Water Purification Systems.

Specifications

Feedwater Source Tap
Flow Rate 6 L/min.
Material Cellulose, Polyester
Pore Size Fine
Purification Method Reverse osmosis
Resin Type Carbon
For Use With (Equipment) WaterPro™ reverse osmosis system
Type Reverse Osmosis Membrane
Weight (Metric) 2.7 kg