missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ WaterPro™ RO Station Dispensing Gun

Catalog No. 16107165
Change view
Click to view available options
Capacity (Metric):
1.0 L/min.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
16107165 1.0 L/min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16107165 Supplier Labconco™ Supplier No. 9113100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with WaterPro™ RO Systems.

Dispensing Gun provides additional means to dispense water from the WaterPro RO System. Trigger permits operation with one hand and provides precise drop-by-drop control or locks to provide a continuous stream of water. Includes gun attached to a 6-foot hose, powder-coated steel gun rest and mounting hardware.

Specifications

Capacity (Metric) 1.0 L/min.
Feed Water Monitoring No
Feedwater Source Tap
Flow Rate 1.0 L/min.
Product Water Type Type 3
TOC Monitor with UV Intensity Monitoring No
Type Dispensing Gun
For Use With (Equipment) WaterPro™ reverse osmosis station
Includes 6 ft. Hose, Epoxy coated Steel gun rest, mounting hardware and instruction