Learn More
Labconco™ Vacuum Controller for CentriVap™ Systems
Catalog No. 1631643
Change view
Click to view available options
For Use With:
Diaphragm vacuum pump
Catalog No. For Use With
1631643 Diaphragm vacuum pump
1 options
Catalog No. 1631643 Supplier Labconco™ Supplier No. 7991000
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For use with Diaphragm Vacuum Pumps only
Provides manual control and monitoring of vacuum levels. Analog gauge shows vacuum from 0–30 in.Hg. At the end of concentration, the control valve may opened to break the vacuum.
Specifications
For Use With Diaphragm vacuum pump
Includes Analog gauge showing 0 to 30 in. Hg and rear inlet and outlet ports
Type Vacuum Controller