missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Vacu-Pass™ Cord and Cable Portal

Catalog No. 3402800
Change view
Click to view available options
Cord and Cable Portal
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
3402800 Cord and Cable Portal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3402800 Supplier Labconco™ Supplier No. 3402800

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Contains blank plug for sealing port during non-use

  • Allows passage of a cord or cable through right-side interior wall to the exterior of a Purifier Logic+ Biological Safety Cabinet
  • Includes set of 4 closed-cell neoprene plugs with holes for cords/cables with diameters of 1/8, 1/4, 3/8 and 1/2 inch
  • Includes a blank plug for sealing port during non-use
  • NSF-approved port does not disrupt airflow or allow leakage

Certifications

NSF-approved

Specifications

Product Type Cord and Cable Portal
For Use With (Equipment) Purifier Axiom Biosafety Cabinets
Size Standard
Amperage NA
Certifications/Compliance NSF
Color Black
Includes 4 closed-cell neoprene plugs with holes for cords/cables with of 0.12, 0.25, 0.37 and 0.5 in. dia., blank plug for sealing the port during non-use
Weight (English) 2 lb.
Weight (Metric) 0.9 kg