missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Trap Insert Canister with Stand for Precise™ Glove Boxes

Catalog No. 16108386
Change view
Click to view available options
Material:
Polypropylene
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
16108386 Polypropylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16108386 Supplier Labconco™ Supplier No. 7815301

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Clear polypropylene canister

  • Accommodates any of the following Chemical Trap Inserts: Acid Trap (Mfr. No. 7814800), Moisture Trap (Mfr. No. 7814900), Radioisotope Trap (Mfr. No. 7815000), Solvent Trap (Mfr. No. 7815200), or Ammonia Trap (Mfr. No. 79956000), each sold separately
  • For use with Precise™ Controlled Atmosphere and Basic Glove Boxes,as well as Protector™ Combination and Controlled Atmosphere Glove Boxes

Specifications

Color Clear
Case Material Polypropylene
Material Polypropylene
Maximum Pressure 1 bar
For Use With (Equipment) 7814800 Acid trap, 7814900 moisture trap, 7815000 Radioisotope trap, 7815200 Solvent trap or 7995600 Ammonia trap insert
Includes Stand
Type Canister
Weight (English) 5 lb.
Weight (Metric) 2.3 kg