Learn More
Labconco™ SpillStopper™ Work Surface for 6 Ft. Protector™Premier™and Protector™XL Hoods
Catalog No. 10369164
Change view
Click to view available options
Includes:
Left Rear
Left Rear, Right Rear
Left Side
Left Side, Right Rear
No Cutouts
Right Rear
Right Side
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
10369164 Left Side
10369163 Right Rear
10369162 Left Rear
10369161 No Cutouts
10369165 Right Side
10369166 Left Rear, Right Rear
10369167 Left Side, Right Rear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 10369164 Supplier Labconco™ Supplier No. 9500620
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Labconco SpillStopper™ Work Surface is dished and contoured to conform to the interior liner to contain spills.
  • Corrosion-resistant epoxy resins molded
  • Front edge with large radius to aerodynamically direct airflow in the hood
  • Pre-drilled holes for venting and plumbing passthroughs provided
  • Some surfaces have cutouts for a surface-mounted service fixture and/or a 6 x 3 in. (15.2 x 7.6cm) cup sink
  • Flat work surfaces may also be coupled with an adjustable height base stand to provide auxiliary workspace
  • Service fixture and cup sink are sold separately
9500622
Not for use with Protector Solvent Storage Cabinets
Specifications
For Use With (Equipment) Protector XL Laboratory Hood
Includes Left Side
Product Type Work Surface
Material Epoxy Resin
For Use With (Application) Laboratory Fume Hood Work