Learn More
Labconco™ SpillStopper™ Work Surface for 6 Ft. Protector™Premier™and Protector™XL Hoods
Labconco SpillStopper™ Work Surface is dished and contoured to conform to the interior liner to contain spills.
Supplier:  Labconco™ 9500610
 View more versions of this product
Catalog No. 10369162
Only null left
Description
Description
  • Corrosion-resistant epoxy resins molded
  • Front edge with large radius to aerodynamically direct airflow in the hood
  • Pre-drilled holes for venting and plumbing passthroughs provided
  • Some surfaces have cutouts for a surface-mounted service fixture and/or a 6 x 3 in. (15.2 x 7.6cm) cup sink
  • Flat work surfaces may also be coupled with an adjustable height base stand to provide auxiliary workspace
  • Service fixture and cup sink are sold separately
9500622
Not for use with Protector Solvent Storage Cabinets
Specifications
Chemical Identifiers
Protector XL Laboratory Hood
Work Surface
Epoxy Resin
Laboratory Fume Hood Work
Specifications
Protector XL Laboratory Hood
Work Surface
Laboratory Fume Hood Work
Left Rear
Epoxy Resin
Product Suggestions
Product Suggestions