Learn More
Labconco™ SpillStopper™ Work Surface for 4 Ft. Protector™ Premier™ Hood
Catalog No. 10369153
Change view
Click to view available options
Cut Style:
Left Rear
Left Rear, Right Rear
Left Side
Left Side, Right Rear
No Cutouts
Right Rear
Right Side
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cut Style
10369153 Left Side, Right Rear
10369148 Left Rear
10369147 No Cutouts
10369149 Right Rear
10369150 Left Side
10369152 Left Rear, Right Rear
10369151 Right Side
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 10369153 Supplier Labconco™ Supplier No. 9500422
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Labconco SpillStopper™ Work Surface is dished and contoured to conform to the interior liner to contain spills.
  • Corrosion-resistant epoxy resins molded
  • Front edge with large radius to aerodynamically direct airflow into hood
  • Pre-drilled holes for venting and plumbing pass-throughs provided
  • Some surfaces with cutouts for surface-mounted service fixture and/or 6 x 3 in. (15.2 x 7.6cm) cup sinkcup sink
  • Flat work surfaces may also be coupled with adjustable height base stand to provide auxiliary workspace
Order Info
Service fixture and cup sink sold separately
Specifications
For Use With (Equipment) Protector XL Laboratory Hood
Includes Left Side, Right Rear
Product Type SpillStopper Work Surface (4 ft.)
Material Epoxy Resin
For Use With (Application) Laboratory Fume Hood Work
Cut Style Left Side, Right Rear