missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Serum Bottle Aluminum Seals, Large
Description
- Fits 5mL, 10mL, 20mL, 30mL, 50mL, 100mL and 125mL serum bottle
Specifications
Specifications
| Color | Silver |
| For Use With (Application) | Fits 5mL, 10mL, 20mL, 30mL, 50mL, 100mL and 125mL Serum bottles |
| Material | Aluminum |
| Sterility | Non-sterile |
| Weight (Metric) | 0.5kg |
| Weight (English) | 1 lb. |
| Thread Size (Metric) | 57 mm |
| Neck Finish | 57 mm |
| Disposable | Single-use |
| Product Type | Cap |
| Show More |