missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Purifier™ Vertical Clean Bench

Catalog No. 16108243
Change view
Click to view available options
Width (English):
24 in.
36 in.
48 in.
Voltage:
115 V
230 V
Plug Type:
No Plug
North America Plug
Includes:
Airflow Monitor
Bench Only
UV, Panel, Timer
UV, Panel, Timer, Airflow Monitor
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English) Voltage Plug Type Includes
16108243 24 in. 115 V North America Plug Bench Only
16108246 24 in. 115 V North America Plug Airflow Monitor
16108245 24 in. 115 V North America Plug UV, Panel, Timer
16108247 24 in. 115 V North America Plug UV, Panel, Timer, Airflow Monitor
16108252 36 in. 230 V North America Plug Bench Only
16108256 36 in. 230 V No Plug Bench Only
16108254 36 in. 230 V North America Plug Airflow Monitor
16108253 36 in. 230 V North America Plug UV, Panel, Timer
16108255 36 in. 230 V North America Plug UV, Panel, Timer, Airflow Monitor
16108260 48 in. 115 V North America Plug Bench Only
16108261 48 in. 115 V North America Plug UV, Panel, Timer
16108263 48 in. 115 V North America Plug UV, Panel, Timer, Airflow Monitor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Purifier Vertical Clean Benches do not provide user protection. Biohazardous materials, toxins and radionuclides should be handled in Class I or Class II biological safety cabinets. (See Purifier Class I Safety Enclosures and Delta Series Cabinets).
Catalog No. 16108243 Supplier Labconco™ Supplier No. 3970200

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Purifier Vertical Clean Benches do not provide user protection. Biohazardous materials, toxins and radionuclides should be handled in Class I or Class II biological safety cabinets. (See Purifier Class I Safety Enclosures and Delta Series Cabinets).

Requires

Work surface and base stand (sold separately).

Protect work from particulate contamination by choosing a Purifier™ Vertical Clean Bench.

  • Nominal downflow velocity of 45-65 fpm and aerodynamic air foil
  • Supply HEPA filter, 99.99% efficient on particles 0.3 Micron
  • ISO Class 5 air (formerly Class 100 per Federal Standard 209E)
  • Variable speed built-in blower with motorized impeller and solid state control rated for 10 A
  • Intrinsically-sound, negative pressure design
  • Minihelic pressure gauge and disposable prefilter
  • UV-resistant, glacier white and gray, powder-coated aluminum and steel construction
  • UV-opaque and resistant 6.3mm (0.25 in.) thick tempered safety glass sash and sides
  • Light and blower switches
  • Air diffuser and 23.8cm (9.4 in.) high sash opening
  • Ergonomic 10° angled and hinged sash that pivots upward, locks to a loading height of 50.8cm (20 in.) and has wiping seals to promote ISO Class 5 conditions
  • Two utility ports with iris openings and plugs
  • ETL Listing (115 volt, 60 Hz models)

Order Info

Vertical Clean Benches can be ordered with bifold doors; specify when ordering. Guardian™ Airflow Monitor is offered as factory-installed option on all benches; specify when ordering.

Warranty

One year parts and service

Specifications

Product Type Clean Bench
Coating Powder
Filter Type HEPA
Lighting Type Fluorescent
Material Powder-coated Aluminum and Steel
Amperage 5 A
Certifications/Compliance CAN/CSA C22.2, UL, CE
Color Glacier White, Gray
Depth (English) 28.6 in.
Depth (English) Interior 25.3 in.
Depth (Metric) 72.64 cm
Depth (Metric) Interior 64.3 cm
For Use With (Application) Plant tissue culture, media plate preparation, electronics inspection, medical device assembly and pharmacy drug preparation
Height (English) 36.8 in.
Height (English) Interior 22.7 in.
Height (Metric) 93.47 cm
Height (Metric) Interior 57.7 cm
Hertz 50/60 Hz
Includes Bench Only
IP Rating None
Plug Type North America Plug
Ports Utility
Voltage 115 V
Weight (English) 125 lb.
Weight (Metric) 57 kg
Width (English) 24 in.
Width (English) Interior 22.8 in.
Width (Metric) 60.96 cm
Width (Metric) Interior 57.9 cm
Show More Show Less