Learn More
Labconco™ Pure-Vu™ Microscope Seals
Microscope seals for use with Labconco Purifier Cell Logic+ Biosafety Cabinets.
Supplier:  Labconco™ 4027810
 View more versions of this product
Catalog No. 16-317-146
Only null left
Description
Description
Microscope seals for use with Purifier Cell Logic+ Biosafety Cabinets. Universal seals are clear blank membranes which may be cut with 0.5 in. diameter holes or punched with Pure-Vu Punch Kit 4027801 to fit your microscope’s eyepieces. Convenient, pre-punched seals include 0.5 in. diameter holes at set heights. Given height indicates distance from base of microscope to centerline of eyepiece.
Specifications
Specifications
Universal Pure-Vu XL Seal
Purifier cell logic biosafety cabinet with scope ready package
0.5 lb.
0.2 kg
Product Suggestions
Product Suggestions