missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Filtered Fume Hood Accessory, HEPA Filter
For use with Protector Echo and Airo Filtered Fume Hood
Supplier: Labconco™ 9577401
Description
- Optional HEPA filter can be placed between prefilter and main level Neutrodine filter to provide capture of non-hazardous particulates
- Efficiency: 99.995%, H14 type
- Number of filters needed depends on hood's width
Order Info
Quantity required per model: 3 ft. Hood- 1 in.; 4 ft. Hood- 2 in.; 5 ft. Hood- 3 in.; 6 ft. Hood- 4 in.; 8 ft. Hood- 5 in.
Warranty
One year
Specifications
| HEPA Filter | |
| HEPA | |
| 76.2 cm | |
| Protector™ Echo™ and Airo™ Filtered Fume Hood | |
| 10.1 cm | |
| 16 in. |
| Powder | |
| 30 in. | |
| 16 x 30 x 3.5 in. | |
| 4 in. | |
| No services | |
| 40.6 cm |