missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Protector™ Double Stainless Steel Combination Glove Boxes

Catalog No. 16200456
Change view
Click to view available options
Certifications/Compliance:
UL, ETL, CAN/CSA
Plug Type:
North America, 100/115 V
North America, 208/230 V
Voltage:
100/115 V
208/230 V
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance Plug Type Voltage
16200456 UL, ETL, CAN/CSA North America, 100/115 V 100/115 V
16200460 UL, ETL, CAN/CSA North America, 208/230 V 208/230 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16200456 Supplier Labconco™ Supplier No. 5080162

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Auto-Pressure Controller

Has the capability to function as either a filtered box or a controlled atmosphere glove box

  • Inner and outer pivoting transfer chamber doors, counterbalanced and equipped with quick-latches
  • Two rear-mounted inlet and outlet 0.75 in. OD compression fittings for connection to optional AtmosPure™ Re-Gen Gas Purifier or other accessory drying train
  • Interior right-hand side wall-mounted electrical receptacle (duplex on 100/115V models) with 10A maximum
  • Interior left-hand side wall-mounted pass-through with plug
  • Exceed Class 1 atmosphere containment conditions for oxygen permeation <1.67ppm/min per ISO 10648-2 test method
  • Centrally-mounted electrical receptacle
  • Type 304 stainless steel liner, Type 304 stainless steel inner and outer transfer chamber doors and Type 304 stainless steel front panel
  • Four fluorescent lights with two viewing windows and Four neoprene gloves and glove ports
  • Illumination exceeds 80 footcandles
  • Work with both atmosphere-sensitive and/or hazardous materials.

Specifications

Product Type Glove Box
Filter Type ULPA
Lighting Type Fluorescent (2 x 25 W)
Material Stainless Steel
Amperage 10 A
Certifications/Compliance UL, ETL, CAN/CSA
Depth (English) 34.4 in.
Depth (Metric) 87.4 cm
Display LCD
For Use With (Application) Laboratories that have multiple types of glove box applications or anticipate that their applications may change in the future (atmosphere sensitive materials and hazardous materials)
For Use With (Equipment) AtmosPure™ Re-Gen gas purifier
Height (English) 45.7 in.
Height (Metric) 116.1 cm
Plug Type North America, 100/115 V
Ports 4
Voltage 100/115 V
Weight (English) 1300 lb.
Weight (Metric) 590 kg
Width (English) 108.9 in.
Width (English) Interior 81.8 in.
Width (Metric) 276.6 cm
Width (Metric) Interior 207.77 cm
Show More Show Less