Learn More
Labconco™ Protector™ Acid and Standard Storage Cabinets Shelf Kits
Catalog No. 1630540
Change view
Click to view available options
For Use With (Equipment):
Protector Acid Storage Cabinet
Protector Standard Storage Cabinet
Width (English) Exterior:
24 in.
30 in.
36 in.
48 in.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Width (English) Exterior
1630540 Protector Standard Storage Cabinet 48 in.
1630542 Protector Acid Storage Cabinet 24 in.
1630543 Protector Acid Storage Cabinet 36 in.
1630544 Protector Acid Storage Cabinet 48 in.
1630545 Protector Acid Storage Cabinet 30 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 1630540 Supplier Labconco™ Supplier No. 9919100
Shelf Kits provide additional storage space in Labconco Protector Standard and Acid Base Cabinets.
Interior Shelf Kit provides additional storage space inside Labconco Protector Standard Storage Cabinets. Includes powder-coated steel shelf and mounting hardware. Shelf may be mounted at one of three heights: 13.0 in., 15.25 in., 17.0 in. (9.5 in., 11.25 in. and 13.0 in. for ADA height cabinets).
Specifications
For Use With (Equipment) Protector Standard Storage Cabinet
Width (Metric) Exterior 121.92 cm
Product Type Shelf Kit
Width (English) Exterior 48 in.
Includes Epoxy Coated Steel shelf and mounting hardware
Material Steel