missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ PrimeMate™ Vacuum Pump Oil

Catalog No. p-4893732
Change view
Click to view available options
Material:
Hydro-treated Paraffinic Hydrocarbon Oil
Synthetic Hydrocarbon Oil
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
10269739 Hydro-treated Paraffinic Hydrocarbon Oil
10269740 Synthetic Hydrocarbon Oil
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10269739 Supplier Labconco™ Supplier No. 1988001
Only null left

For use with the Labconco PrimeMate Oil Change System

  • Choice of one gallon (3.8L) hydro-treated paraffinic hydrocarbon oil or synthetic hydrocarbon oil
  • Recommended for Labconco general purpose vacuum pumps

Specifications

Product Type Vacuum Pump Oil
Format Oil
For Use With (Equipment) PrimeMate Oil Change System
Material Hydro-treated Paraffinic Hydrocarbon Oil
Volume (English) 1 gal.
Volume (Metric) 3.8 L