Learn More
Labconco™ Logic+™ 9-Pin and 25-Pin RS232 Connection Kit
Catalog No. 3858311
Change view
Click to view available options
For Use With (Application):
Connecting the purifier logic+ biosafety cabinet to a user supplied data recorder or building monitoring system
Connecting the purifier™ logic™ biosafety cabinet to a user supplied data recorder or building monitoring system
Includes:
Pc board, hardware and cable with 25-pin connection
Pc board, hardware and cable with 9-pin connection
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Includes
3858311 Connecting the purifier™ logic™ biosafety cabinet to a user supplied data recorder or building monitoring system Pc board, hardware and cable with 25-pin connection
3858310 Connecting the purifier logic+ biosafety cabinet to a user supplied data recorder or building monitoring system Pc board, hardware and cable with 9-pin connection
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 3858311 Supplier Labconco™ Supplier No. 3858311
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For use with Biosafety Cabinet, Procedure Station, Clean Bench or Filtered PCR Enclosure
3858310
  • Kit contains PC board, hardware and cable with 9-pin connection for connecting the Purifier Logic+ Biosafety Cabinet to a user-supplied data recorder or building monitoring system
3858311
  • Kit contains PC board, hardware and cable with 25-pin connection for connecting the Purifier Logic+ Biosafety Cabinet to a user-supplied data recorder or building monitoring system
Specifications
Product Type Connection Kit
For Use With (Equipment) Purifier Logic and Logic+ Biosafety Cabinets
Plug Type RS 232 25-pin
For Use With (Application) Connecting the purifier™ logic™ biosafety cabinet to a user supplied data recorder or building monitoring system
Includes Pc board, hardware and cable with 25-pin connection
Weight (English) 1 lb.
Weight (Metric) 0.5 kg