missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ LabSolutions™ Powder Detergent

Catalog No. p-84408
Change view
Click to view available options
Volume (English):
10 lb.
27.5 lb.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
04334 10 lb.
04335 27.5 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 04334 Supplier Labconco™ Supplier No. 4422000
Only null left

Includes

Scoop

Removes grease, blood, agar, protein digestates, and other contaminants

For automatic glassware washers.

  • Nonfoaming powder
  • Free-rinsing and noncorrosive
  • Contains phosphates
  • Approximate pH of 1% solution: 12.5

Specifications

Product Type Detergent
Color-coded Yes
Form Powder
pH 12.5
Scent Unscented
Sterility Non-sterile
Composition Phosphate
For Use With (Application) For use in Glassware washers, removes range of contaminants including Protein digestates, Agar, Blood and Grease
Size 10 lb.
Volume (English) 10 lb.