Learn More
Labconco™ Freeze-Drying Ampule Accessories
Freeze drying ampule accessories allow ampules to be used with Labconco FreeZone Freeze Dryers.
Supplier:  Labconco™ 7593401
 View more versions of this product
Catalog No. 1026980
Only null left
Description
Description
Freeze drying ampule accessories allow ampules to be used with Labconco FreeZone Freeze Dryers.
  • Large rubber split stoppers fit 5, 10, 20, 30, 50, 100 and 125 ml Labconco Serum Bottles (bottles with 20 mm corkage).
    • Package of 100.
  • Oxygen/Natural Gas Sealing Torch is specifically designed for flame sealing freeze dry ampules.
    • Seals all types of heat-resistant glass.
    • Connects to natural gas, butane or propane and oxygen with 1/4 in. ID hose connectors.
  • Ampule valve adapter provides connection of ampules to drying chambers and manifolds by connecting ampules to valve ports or stainless steel stems.
    • Ten per package.
Specifications
Chemical Identifiers
Freeze drying system
Specifications
Ampule Adapter
Freeze drying system
0.2 kg
Neoprene
0.5 lb.
Product Suggestions
Product Suggestions