Learn More
Labconco™ Fast-Freeze™ Flask Adapter
Fast-Freeze™ Flask Adapter for connecting Fast-Freeze™ flasks to freeze-dryer valves.
$35.04 - $148.52
Specifications
| For Use With (Application) | For connecting fast freeze™ flasks to freeze-dryer valves |
|---|---|
| For Use With (Equipment) | Fast freeze™ flask |
| Weight (English) | 0.5 lb. |
| Weight (Metric) | 0.2 kg |
| Catalog Number | Mfr. No. | Diameter (English) | Material | Style | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Diameter (English) | Material | Style | Price | Quantity & Availability | |||
|
1026957A
|
Labconco™
7545000 |
0.5 in. | Borosilicate Glass | Straight |
Each for $35.04
|
|
|||
|
1026958A
|
Labconco™
7545200 |
0.75 in. | Borosilicate Glass | Straight |
Each for $44.45
|
|
|||
|
1026959B
|
Labconco™
7546000 |
0.5 to 0.75 in. | Borosilicate Glass | 45° bend |
Each for $100.04
|
|
|||
|
1026959A
|
Labconco™
7545400 |
0.5 to 0.75 in. | Borosilicate Glass | Straight |
Each for $62.82
|
|
|||
|
1026958B
|
Labconco™
7545800 |
0.75 in. | Borosilicate Glass | 45° bend |
Each for $61.96
|
|
|||
|
1026957B
|
Labconco™
7545600 |
0.5 in. | Borosilicate Glass | 45° bend |
Each for $61.96
|
|
|||
|
1026960A
|
Labconco™
7547000 |
0.5 in. | Stainless Steel | Straight |
Each for $87.48
|
|
|||
|
1026960B
|
Labconco™
7547400 |
0.5 in. | Stainless Steel | 45° bend |
Each for $148.52
|
|
|||
|
1026961A
|
Labconco™
7547200 |
0.75 in. | Stainless Steel | Straight |
Each for $87.48
|
|
|||
|
1026961B
|
Labconco™
7547600 |
0.75 in. | Stainless Steel | 45° bend |
Each for $113.98
|
|
|||
Description
For connecting Fast-Freeze™ flasks to freeze-dryer valves
- 40 and 80 mL flasks take size 0.5 in. or 0.5 to 0.75 in.
- Larger flasks fit size 0.75 in. or large end of 0.5 to 0.75 in.
Specifications
| For connecting fast freeze™ flasks to freeze-dryer valves | |
| Fast freeze™ flask | |
| 0.5 lb. | |
| 0.2 kg |