Learn More
Labconco™ Drying Chamber for FreeZone™ Freeze-Dry Systems
Drying Chamber for use on FreeZone™ Freeze-Dry Systems. 12-port for 1, 2.5, 6, 12, and 18 L models; 16-port for 6, 12, and 18 L models.
$5,392.61 - $7,354.36
Specifications
Type Drying Chamber
No. of Shelves 0
Material Stainless Steel
For Use With (Application) To provide a means to attach flasks to freeze dry system or contain samples in bulk
View More Specs
Products 2
Catalog Number Mfr. No. No. of Ports Diameter (English) Price Quantity & Availability  
Catalog Number Mfr. No. No. of Ports Diameter (English) Price Quantity & Availability  
1027320
View Documents Promotion Details
Labconco™
7522900
16 13 in.
Each for $7,354.36
Only null left
1027319
View Documents Promotion Details
Labconco™
7522800
12 8 in.
Each for $5,392.61
Only null left
Description
Description
For use on FreeZone™ Freeze-Dry Systems: 1, 2.5, 4.5, 6, 8, 12 or 18 Liter models
  • Place small sample inside freeze-dry flasks and attach to valves
  • Type 304 stainless-steel construction
  • Weld-free for dependable vacuum operation
  • Neoprene valves accommodate both 0.5 and 0.75 in. adapters, gaskets and clear acrylic top for complete visibility during bulk drying
  • Compatible with FreeZone 1, 2.5, 4.5, 6, 8, 12 or 18 Liter Freeze Dryers
Specifications
Specifications
Drying Chamber
0
Stainless Steel
To provide a means to attach flasks to freeze dry system or contain samples in bulk