Learn More
Labconco™ DNA Rotor for CentriVap™ System
Catalog No. 1631771
Change view
Click to view available options
Holds:
56 Autosampler Vials
66 Autosampler Vials
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
1631771 56 Autosampler Vials
1631770 66 Autosampler Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 1631771 Supplier Labconco™ Supplier No. 7824702
Acid-Resistant Thermoplastic Rotors hold tubes at an angle to maximize surface area.
Acid-Resistant Thermoplastic Rotors hold tubes at an angle to maximize surface area. They are easily interchangeable. Sample containers are not included. For use with Labconco CentriVap Vacuum Concentrators.
Specifications
Material Thermoplastic
Type DNA Rotor
For Use With (Equipment) CentriVap™ system
Holds 56 Autosampler Vials
Shipping Weight (English) 3 lb.
Shipping Weight (Metric) 1.4 kg
Weight (English) 3 lb.
Weight (Metric) 1.4 kg