missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Clear Stoppering Chamber
Clear Stoppering Chamber for use with 6 L, 12 L and 18 L Freezone™ Dryers.
$15406.00 - $15406.00
Specifications
| Type | Clear Stoppering Chamber |
|---|---|
| No. of Shelves | 1 |
| Temperature Range (Metric) | Up to 40°C |
| Material | Acrylic |
| For Use With (Application) | To provide economical means of stoppering serum bottles |
| Catalog Number | Mfr. No. | Electrical Requirements | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Electrical Requirements | Price | Quantity | |||||
|
10-030-164
|
Labconco™
7868020 |
115 V, 50/60 Hz, Domestic |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Allows flasks attachment with freeze dry system or contain samples in bulk
Specifications
| Clear Stoppering Chamber | |
| Up to 40°C | |
| To provide economical means of stoppering serum bottles | |
| 13.5 in. | |
| 23 in. | |
| 50/60 Hz | |
| 13.6 kg |
| 1 | |
| Acrylic | |
| 6 L, 12 L and 18 L FreeZone™ Dryers | |
| 34 cm | |
| 58.4 cm | |
| 30 lb. |