missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Canisters with Stands

Catalog No. 1631612
Change view
Click to view available options
Material:
Polypropylene
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
1631612 Polypropylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1631612 Supplier Labconco™ Supplier No. 7815300

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Clear polypropylene canisters allow for visual inspection of trap contents

  • Canister has inlet and outlet 1.9cm (0.75 in.) NPT, 1.3cm (0.5 in.) tube fittings
  • Weight: 2.3kg (5 lb.)
Accommodate any of the following trap inserts (sold separately):
  • Acid Trap Insert
  • Moisture Trap Insert
  • Radioisotope Trap Insert
  • Solvent Trap Insert
  • Ammonia Trap Insert

Specifications

Color Clear
Material Polypropylene
For Use With (Equipment) RapidVap™ vacuum evaporation system
Type Canister With Stand
Weight (English) 5 lb.
Weight (Metric) 2.3 kg