missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Benchtop Fume Adsorbers-2

Catalog No. 10090102
Change view
Click to view available options
Filter Type:
Organic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Filter Type
10090102 Organic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10090102 Supplier Labconco™ Supplier No. 3955400

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed for light-duty applications involving organic solvents, formaldehyde, ammonia or low molecular weight amines

  • Two built-in motorized impellers to provide fast velocity up to 35fp
  • Glacier white, dry powder, epoxy-coated steel housing with perforated baffle for uniform airflow
  • Clear, one-piece, 3/8 in. thick, shatter-resistant acrylic canopy
  • Quiet operation at 58-63 dBA
  • Pre-wired with rear-mounted auxiliary electrical receptacle and top-mounted blower switch with circuit breaker
  • Rear-mounted air sampling port
  • Exterior dimensions: 15.4 x 23.4 x 60 in. (H x D x W) (39.1 x 59.4 x 152.4cm)
  • Interior dimensions: 13.5 x 14.5 in. (H x D) (34.2 x 36.8cm)

Certifications

All models conform to the following regulations and standards:

  • UL 3101-1/61010-1 (115V models)
  • CAN/CSA C22.2 No. 1010.0 (115V models)
  • CE Conformity Marking (230V models)

Specifications

Product Type Fume Adsorber
Filter Type Organic
Amperage 2 A
Certifications/Compliance CAN/CSA C22.2, UL
Depth (English) 23.4 in.
Depth (English) Interior 14.5 in.
Depth (Metric) 59 cm
Depth (Metric) Interior 36.8 cm
For Use With (Application) Designed for light-duty applications involving organic solvents, formaldehyde, ammonia or low molecular weight amines
Height (English) 15.4 in.
Height (English) Interior 13.5 in.
Height (Metric) 39 cm
Height (Metric) Interior 34.2 cm
Voltage 115 V
Weight (English) 135 lb.
Weight (Metric) 61 kg
Width (English) 60 in.
Width (Metric) 152 cm
Frequency 60 Hz
Show More Show Less