missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ UltraVision™ DAB Away™

Catalog No. TA250DA
Change view
Click to view available options
Volume (Metric):
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
TA250DA 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TA250DA Supplier Epredia™ Supplier No. TA250DA

Remove stains and diaminobenzidine precipitate from your Epredia™ Autostainer using Epredia™ UltraVision™ DAB Away™. Apply to stained glassware, staining pads, and more.

Includes:
  • DAB Away Reagent 1 (125mL)
  • DAB Away Reagent 2 (125mL)
  • DAB Away Decolorizer (250mL)

Specifications

Content And Storage Room temperature. Protect from light.
Volume (Metric) 250 mL