missing translation for 'onlineSavingsMsg'
Learn More

Eagle™ Lab Pack Open Head Poly Drum with Bung Holes, 55 gal.
GREENER_CHOICE

Drum is constructed of blow-molded high-density polyethylene (HDPE) with UV inhibitors, is durable and lightweight, as well as weather and chemical resistant. Bung lid includes two holes (3/4 in. and 2 in. diameters) and is ideal for safe storage, as well as handling or transportation of hazardous materials.

Supplier:  Eagle™ 1655MBBG

 View more versions of this product

Catalog No. 19338407


Only null left

Description

Description

  • Constructed from blow-molded high-density polyethylene (HDPE) with UV inhibitors
  • For use with smaller non-damaged bottles, cans, carboys and 5 gal. pails of non-leaking hazardous materials
  • Bung lid includes two holes, one of each diameter: 0.75 in. and 2 in.
  • If used with leaking materials, drums must be packed with sufficient sorbents
  • No free liquids
Specifications

Chemical Identifiers

Drum
55 gal.
Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration
Open
98 cm
23.5 lb.
Yes
208.45 L
HDPE
38.6 in.
0.75 in., 2 in. (Bung Holes)

Specifications

Drum
Metal Lever-lock
208.45 L
Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration
Open
98 cm
23.5 lb.
Yes
55 gal.
Blue
HDPE
38.6 in.
0.75 in., 2 in. (Bung Holes)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions