missing translation for 'onlineSavingsMsg'
Learn More

Eagle™ Lab Pack Open Head Poly Drum with Bung Holes, 30 gal.
GREENER_CHOICE

Catalog No. 19338451
Change view
Click to view available options
Color:
Black
Blue
Closure Type:
Metal Lever-lock
Plastic Lever-lock
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Closure Type
19338451 Black Metal Lever-lock
19338450 Blue Metal Lever-lock
19338449 Black Plastic Lever-lock
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19338451 Supplier Eagle™ Supplier No. 1601MBLKBG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Drum is constructed of blow-molded high-density polyethylene (HDPE) with UV inhibitors, is durable and lightweight, as well as weather and chemical resistant. Bung lid includes two holes (3/4 in. and 2 in. diameters) and is ideal for safe storage, as well as handling or transportation of hazardous materials.

  • Constructed from blow-molded high-density polyethylene (HDPE) with UV inhibitors
  • For use with smaller non-damaged bottles, cans, carboys and 5 gal. pails of non-leaking hazardous materials
  • Bung lid includes two holes, one of each diameter: 0.75 in. and 2 in.
  • If used with leaking materials, drums must be packed with sufficient sorbents
  • No free liquids

Specifications

Product Type Drum
Closure Included Yes
Closure Type Metal Lever-lock
Capacity (English) 30 gal.
Capacity (Metric) 113.7 L
Color Black
For Use With (Application) Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration
Material HDPE
Head Style Open
Height (English) 26.38 in.
Height (Metric) 67 cm
Diameter (English) Opening 0.75 in., 2 in. (Bung Holes)
Weight (English) 18 lb.
Show More Show Less