missing translation for 'onlineSavingsMsg'
Learn More

Basco Lab Pack Open Head Poly Drum with Bung Holes

Catalog No. 19120090
Change view
Click to view available options
Capacity (English):
30 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19120090 30 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19120090 Supplier Basco Supplier No. 1556BLK
Only null left

Drum is constructed of blow-molded high-density polyethylene (HDPE) with UV inhibitors, is durable and lightweight, as well as weather and chemical resistant.

  • Constructed from blow-molded high-density polyethylene (HDPE) with UV inhibitors
  • For use with smaller non-damaged bottles, cans, carboys and 5 gal. pails of non-leaking hazardous materials
  • Bung lid includes two holes, one of each diameter: 0.75 in. and 2 in.
  • If used with leaking materials, drums must be packed with sufficient sorbents
  • No free liquids

Specifications

Closure Included Yes
Product Type Drum
Closure Type Plastic Lever-lock
Capacity (English) 30 gal.
Capacity (Metric) 113.7 L
Color Black
For Use With (Application) Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration
Material HDPE
Head Style Open
Height (English) 26.38 in.
Height (Metric) 67 cm
Diameter (English) Opening 0.75 in., 2 in. (Bung Holes)
Weight (English) 18 lb.
Show More Show Less