missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eagle™ Lab Pack Open Head Poly Drum, 55 gal.

Drum is constructed of blow-molded high-density polyethylene (HDPE) with UV inhibitors, is durable and lightweight, as well as weather and chemical resistant. Ideal for safe storage, handling or transportation of hazardous materials, spill containment, and bio waste disposal.
Supplier: Eagle™ 1655M
Description
- Constructed from blow-molded high-density polyethylene (HDPE) with UV inhibitors
- For use with smaller non-damaged bottles, cans, carboys and 5 gal. pails of non-leaking hazardous materials
- If used with leaking materials, drums must be packed with sufficient sorbents
- No free liquids
Chemical Identifiers
| Drum | |
| 55 gal. | |
| Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration | |
| Open | |
| 98 cm | |
| 23.5 lb. |
| Yes | |
| 208.45 L | |
| HDPE | |
| 38.6 in. | |
| 23.75 in. (Top), 19.4 in. (Bottom) |
Specifications
| Drum | |
| Metal Lever-lock | |
| 208.45 L | |
| Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration | |
| Open | |
| 98 cm | |
| 23.5 lb. |
| Yes | |
| 55 gal. | |
| Yellow | |
| HDPE | |
| 38.6 in. | |
| 23.75 in. (Top), 19.4 in. (Bottom) |