missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eagle™ Lab Pack Open Head Poly Drum with 2 in. Bung Holes, 30 gal.

Drum is constructed of blow-molded high-density polyethylene (HDPE) with UV inhibitors, is durable and lightweight, as well as weather and chemical resistant. Bung lid includes two, 2 in. holes and is ideal for safe storage, as well as handling or transportation of hazardous materials.
Supplier: Eagle™ 1601MBBRBG2
Description
- Constructed from blow-molded high-density polyethylene (HDPE) with UV inhibitors
- For use with smaller non-damaged bottles, cans, carboys and 5 gal. pails of non-leaking hazardous materials
- Bung lid includes two, 2 in. holes
- If used with leaking materials, drums must be packed with sufficient sorbents
- No free liquids
Chemical Identifiers
| Drum | |
| 30 gal. | |
| Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration | |
| Open | |
| 67 cm | |
| 18 lb. |
| Yes | |
| 113.7 L | |
| HDPE | |
| 26.38 in. | |
| 2 in. (Bung Holes) |
Specifications
| Drum | |
| Metal Bolt Ring | |
| 113.7 L | |
| Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration | |
| Open | |
| 67 cm | |
| 18 lb. |
| Yes | |
| 30 gal. | |
| Blue | |
| HDPE | |
| 26.38 in. | |
| 2 in. (Bung Holes) |