missing translation for 'onlineSavingsMsg'
Learn More

Eagle™ Lab Pack Open Head Poly Drum, 10 gal.
GREENER_CHOICE

Catalog No. p-7222713
Change view
Click to view available options
Color:
Black
Blue
Yellow
Closure Type:
Metal Bolt Ring
Metal Lever-lock
Plastic Lever-lock
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Closure Type
19338417 Black Plastic Lever-lock
19338418 Blue Plastic Lever-lock
19338416 Yellow Plastic Lever-lock
19338421 Blue Metal Bolt Ring
19338420 Blue Metal Lever-lock
19338419 Yellow Metal Lever-lock
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19338417 Supplier Eagle™ Supplier No. 1609BLK
Only null left

Drum is constructed of blow-molded high-density polyethylene (HDPE) with UV inhibitors, is durable and lightweight, as well as weather and chemical resistant. Ideal for safe storage, handling or transportation of hazardous materials, spill containment, and bio waste disposal.

  • Drum is constructed of durable HDPE
  • Nestable storage to optimize shipping and storage space
  • Ideal for use with smaller non-damaged bottles, cans, carboys and non-leaking hazardous materials
  • Available with various closure types including plastic or metal lever locking bands, or bolt rings
  • Food grade quality, FDA 21 CFR 177.1520
  • Operating Temp range (-30°F to 130°F)
  • Drop tested-max weight @ 0°F (48 hr.)
  • Flammability Rating: UL 94HB
  • If used with leaking containers, must pack with sufficient sorbents
  • No free liquids

Specifications

Product Type Drum
Closure Included Yes
Closure Type Plastic Lever-lock
Capacity (English) 10 gal.
Capacity (Metric) 37.9 L
Color Black
For Use With (Application) Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration
Material HDPE
Head Style Open
Max. Temperature (Metric) 54.4°C
Height (English) 18 in.
Height (Metric) 45.7 cm
Weight (English) 7.5 lb.
Show More Show Less