Research Products Corporation RPI Color Coded Multi-Purpose Laboratory Tape, 1 Inch Core, 1 Inch Wide, 500 Inches per Roll, Green
Catalog No. 50998720
Laboratory tape adheres to any clean dry surface including glass, plastic, metal, paper, or rubber and removes without leaving a sticky residue. Can be marked with pencil, pen or solvent ink markers. Tape adhesive is oilproof, waterproof and acid resistant.
Catalog No.
50998720
Supplier
Research Products CorporationSupplier No.
RPI 560101G
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Laboratory tape adheres to any clean dry surface including glass, plastic, metal, paper, or rubber and removes without leaving a sticky residue. Can be marked with pencil, pen or solvent ink markers. Tape adhesive is oilproof, waterproof and acid resistant.