Learn More
Fisherbrand™ Lab Grade Cylindrical Dewar Flask with Aluminum Housing and Handle
Borosilicate glass with silvered double walls and a convenient handle
$835.54 - $2,594.96
Specifications
| Material | Aluminum |
|---|---|
| For Use With (Application) | Liquid Nitrogen |
| Includes | Handle |
Description
- Cylindrical- Wide Mouth
- Borosilicate Glass
- Silvered Double Walls
- PE Stopper Included
- For Use with Liquified Gases
- Aluminum Housing
- Lab Grade
- Coated Metal Handle
Specifications
| Aluminum | |
| Liquid Nitrogen | |
| Handle |