missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Disposable Lab Coats
Single-use lab coats which include front snap closure, and wrist cuffs for a comfortable and secure fit.
Supplier: Fisherbrand™ 128930063B
Description
- Fabric: 40 g/m2 spunbond meltblown spunbond (SMS) fabric
- Knit cuff
- Three pockets
- Single use
- Front snap closure
Specifications
| General Purpose Lab Coat | |
| Unisex | |
| Snaps | |
| Single-use | |
| 10 packs of 10 |
| Large | |
| SMS | |
| Knit | |
| Non-sterile |