missing translation for 'onlineSavingsMsg'
Learn More

Elma™ Lab Clean N10 Ultrasonic Cleaning Solution

Catalog No. 08000157
Change view
Click to view available options
Volume (Metric):
2.5 L
10 L
Quantity:
1 Ea.
4 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Quantity
08000157 10 L 1 Ea.
08000156 2.5 L 4 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Irritating to eyes.
Catalog No. 08000157 Supplier Elma™ Supplier No. 800 0077

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Irritating to eyes.

Safely cleans sensitive materials in an ultrasonic bath or automatic washer. Tovatech Elma Lab Clean N10 Cleaning Solution removes emulsions, marking and label residues, light grease and oil, fingerprintes and dust.

  • Neutral pH general-purpose formulation
  • Ideal for gentle cleaning of aluminum and light metals as well as any other material
  • Residue-free - Leaves no residue after rinsing with DI water
  • Dilute with water to 2% for use in an ultrasonic cleaner or to 1% for a labware washer
  • Dilute with either tap water or deionized water
  • Biodegradable

Applications

  • For cleaning any sensitive metal, plastic, glass, or ceramic surface
  • Cleans laboratory sieves, aluminum surfaces prior to anodizing or coating and laser machined parts

Specifications

Product Type Ultrasonic Cleaning Solution
Form Liquid
pH Neutral
Content And Storage Keep between 43°F to 85°F, shelf life 2 years
For Use With (Application) Cleaning Any Sensitive Metal, Plastic, Glass or Ceramic Surface
Quantity 1 Ea.
Volume (Metric) 10 L