missing translation for 'onlineSavingsMsg'
Learn More

L-Valine methyl ester hydrochloride, 99%

Catalog No. AAA1589209
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1589209 10 g
AAA1589218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1589209 Supplier Thermo Scientific Chemicals Supplier No. A1589209
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6306-52-1 | C6H13ClNO2 | 166.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6306-52-1
Molecular Formula C6H13ClNO2
Molecular Weight (g/mol) 166.63
MDL Number MFCD00012497
InChI Key RFMIEHOYULVMNZ-UHFFFAOYNA-N
Synonym l-valine methyl ester hydrochloride, h-val-ome.hcl, methyl valinate hydrochloride, s-methyl 2-amino-3-methylbutanoate hydrochloride, h-l-val-ome hcl, methyl l-valinate hydrochloride, valine methyl ester hydrochloride, s-2-amino-3-methyl-butyric acid methyl ester hydrochloride, h-val-ome hcl, methyl 2s-2-amino-3-methylbutanoate hydrochloride
PubChem CID 111190
IUPAC Name methyl (2S)-2-amino-3-methylbutanoate;hydrochloride
SMILES [Cl].COC(=O)C(N)C(C)C

Specifications

Melting Point 166°C (decomposition)
Beilstein 3594960
Quantity 10 g
Formula Weight 167.64
Optical Rotation +16° (c=2 in Water)
Percent Purity 99%
Chemical Name or Material L-Valine methyl ester hydrochloride
EINECSNumber 228-620-9
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only