missing translation for 'onlineSavingsMsg'
Learn More

L-Valine Ethyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. V01425G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V01425G 5 g
V014225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V01425G Supplier TCI America Supplier No. V01425G

Chemical Identifiers

CAS 17609-47-1
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00012511
InChI Key PQGVTLQEKCJXKF-RGMNGODLSA-N
Synonym l-valine ethyl ester hydrochloride, h-val-oet.hcl, ethyl l-valinate hydrochloride, h-val-oet hcl, valine ethyl ester hydrochloride, h-l-meval-ome*hcl, valine, ethyl ester, hydrochloride, ethyl 2s-2-amino-3-methylbutanoate hydrochloride, l-ethyl 2-amino-3-methylbutanoate hydrochloride, ethyl l-valinate hcl
PubChem CID 87181
IUPAC Name ethyl (2S)-2-amino-3-methylbutanoate hydrochloride
SMILES Cl.CCOC(=O)[C@@H](N)C(C)C

Specifications

Melting Point 104°C
Color White
Quantity 5 g
Formula Weight 181.66
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Valine Ethyl Ester Hydrochloride