missing translation for 'onlineSavingsMsg'
Learn More

L-Valine ethyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog No. AAH5946606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5946606 5 g
AAH5946603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5946606 Supplier Thermo Scientific Chemicals Supplier No. H5946606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17609-47-1
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00012511
InChI Key PQGVTLQEKCJXKF-RGMNGODLSA-N
Synonym l-valine ethyl ester hydrochloride, h-val-oet.hcl, ethyl l-valinate hydrochloride, h-val-oet hcl, valine ethyl ester hydrochloride, h-l-meval-ome*hcl, valine, ethyl ester, hydrochloride, ethyl 2s-2-amino-3-methylbutanoate hydrochloride, l-ethyl 2-amino-3-methylbutanoate hydrochloride, ethyl l-valinate hcl
PubChem CID 87181
SMILES Cl.CCOC(=O)[C@@H](N)C(C)C

Specifications

Melting Point 102°C to 105°C
Linear Formula (CH3)2CHCH(NH2)CO2C2H5·HCl
IUPAC Name ethyl (2S)-2-amino-3-methylbutanoate hydrochloride
Quantity 5 g
Formula Weight 181.66
Optical Rotation +6.7° (c=2 in H2O)
Percent Purity 98%
Chemical Name or Material L-Valine ethyl ester hydrochloride
EINECSNumber 241-580-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only