missing translation for 'onlineSavingsMsg'
Learn More

L-Tyrosine, MP Biomedicals™

Catalog No. p-4404571
Change view
Click to view available options
Quantity:
50 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10318350 50 g
ICN10318391 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10318350 Supplier MP Biomedicals Inc Supplier No. 0210318350

Chemical Identifiers

CAS 60-18-4
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD00002606
InChI Key OUYCCCASQSFEME-SVGMAFHSNA-N
Synonym l-tyrosine, tyrosine, s-tyrosine, p-tyrosine, h-tyr-oh, l-p-tyrosine, 2s-2-amino-3-4-hydroxyphenyl propanoic acid, 4-hydroxy-l-phenylalanine, l---tyrosine, tyrosine, l
PubChem CID 6057
ChEBI CHEBI:17895
IUPAC Name (2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(O)C=C1)C(O)=O

Specifications

Color White
Assay Percent Range 0.97
Density 1.4g/mL
Quantity 50 g
Formula Weight 181.19
Physical Form Powder
Chemical Name or Material L-Tyrosine
EINECSNumber 200-460-4
Recommended Storage Store at Room Temperature (15 to 30°C).