missing translation for 'onlineSavingsMsg'
Learn More

L-Tyrosine Methyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. p-7136613
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11085G 5 g
T110825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T11085G Supplier TCI America Supplier No. T11085G
Only null left

Chemical Identifiers

CAS 3417-91-2
Molecular Formula C10H14ClNO3
Molecular Weight (g/mol) 231.68
MDL Number MFCD00012607
InChI Key VXYFARNRGZWHTJ-MVRPGTNWNA-N
Synonym l-tyrosine methyl ester hydrochloride, h-tyr-ome.hcl, methyl l-tyrosinate hydrochloride, h-tyr-ome hcl, methyl tyrosinate hydrochloride, unii-w42m0mi271, tyrosine methyl ester hydrochloride, methyl l-tyrosinate hcl, s-methyl 2-amino-3-4-hydroxyphenyl propanoate hydrochloride, s-2-amino-3-4-hydroxy-phenyl-propionic acid methyl ester hydrochloride
PubChem CID 76956
IUPAC Name methyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate hydrochloride
SMILES Cl.COC(=O)[C@@H](N)CC1=CC=C(O)C=C1

Specifications

Color White
Quantity 5 g
Formula Weight 231.68
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Tyrosine Methyl Ester Hydrochloride