missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Tyrosine [Free Base], MP Biomedicals
$123.68 - $664.53
Chemical Identifiers
| CAS | 60-18-4 |
|---|---|
| Molecular Formula | C9H11NO3 |
| Molecular Weight (g/mol) | 181.19 |
| MDL Number | MFCD00002606 |
| InChI Key | OUYCCCASQSFEME-SVGMAFHSNA-N |
| Synonym | l-tyrosine, tyrosine, s-tyrosine, p-tyrosine, h-tyr-oh, l-p-tyrosine, 2s-2-amino-3-4-hydroxyphenyl propanoic acid, 4-hydroxy-l-phenylalanine, l---tyrosine, tyrosine, l |
| PubChem CID | 6057 |
| ChEBI | CHEBI:17895 |
| IUPAC Name | (2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid |
| SMILES | N[C@@H](CC1=CC=C(O)C=C1)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
ICN19475950
|
N/A | 50 g |
Each for $123.68
|
|
|||||
|
ICN19475980
|
N/A | 100 g |
Each for $212.31
|
|
|||||
|
ICN19475990
|
N/A | 500 g |
Each for $664.53
|
|
|||||
Chemical Identifiers
| C9H11NO3 | |
| MFCD00002606 | |
| l-tyrosine, tyrosine, s-tyrosine, p-tyrosine, h-tyr-oh, l-p-tyrosine, 2s-2-amino-3-4-hydroxyphenyl propanoic acid, 4-hydroxy-l-phenylalanine, l---tyrosine, tyrosine, l | |
| CHEBI:17895 | |
| N[C@@H](CC1=CC=C(O)C=C1)C(O)=O |
Specifications
| 60-18-4 | |
| MFCD00002606 | |
| OUYCCCASQSFEME-SVGMAFHSNA-N | |
| (2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid | |
| 6057 | |
| 181.2 | |
| Crystals |
| C9H11NO3 | |
| l-tyrosine, tyrosine, s-tyrosine, p-tyrosine, h-tyr-oh, l-p-tyrosine, 2s-2-amino-3-4-hydroxyphenyl propanoic acid, 4-hydroxy-l-phenylalanine, l---tyrosine, tyrosine, l | |
| N[C@@H](CC1=CC=C(O)C=C1)C(O)=O | |
| 181.19 | |
| CHEBI:17895 | |
| Cell Culture | |
| L-Tyrosine [Free Base] |
Safety and Handling
Recommended Storage : Room temperature