missing translation for 'onlineSavingsMsg'
Learn More

L-Tyrosine, FCC, 98.5-101.5%, Spectrum™ Chemical

Catalog No. p-7157854
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
Packaging:
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603988 25 g Poly Bottle
18603985 100 g Poly Bottle
18603986 1 kg Poly Pail
18603987 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18603988 Supplier Spectrum Chemical Mfg Cor Supplier No. T115025GM
Only null left

T1150, 60-18-4, C9H11NO3

Spectrum™ Chemical L-Tyrosine, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 60-18-4
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD00002606
InChI Key OUYCCCASQSFEME-SVGMAFHSNA-N
IUPAC Name (2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(O)C=C1)C(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.001
Loss on Drying 0.003
Packaging Poly Bottle
Quantity 25 g
Optical Rotation -10.0° to -11.0°
Percent Purity 98.5 to 101.5%
Grade FCC