missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Tyrosine Ethyl Ester 98.0+%, TCI America™
$57.75 - $229.10
Chemical Identifiers
| CAS | 949-67-7 |
|---|---|
| Molecular Formula | C11H15NO3 |
| Molecular Weight (g/mol) | 209.25 |
| MDL Number | MFCD00063046 |
| InChI Key | SBBWEQLNKVHYCX-JTQLQIEISA-N |
| Synonym | l-tyrosine ethyl ester, ethyl l-tyrosinate, tyrosine ethyl ester, h-tyr-oet, l-tyrosine, ethyl ester, tyr-oet, ethyl tyrosine ester, l-tyrosine ethyl, unii-17rpw76a9g, s-ethyl 2-amino-3-4-hydroxyphenyl propanoate |
| PubChem CID | 70364 |
| ChEBI | CHEBI:9798 |
| IUPAC Name | ethyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate |
| SMILES | CCOC(=O)[C@@H](N)CC1=CC=C(O)C=C1 |
Chemical Identifiers
| 949-67-7 | |
| 209.25 | |
| SBBWEQLNKVHYCX-JTQLQIEISA-N | |
| 70364 | |
| ethyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate |
| C11H15NO3 | |
| MFCD00063046 | |
| l-tyrosine ethyl ester, ethyl l-tyrosinate, tyrosine ethyl ester, h-tyr-oet, l-tyrosine, ethyl ester, tyr-oet, ethyl tyrosine ester, l-tyrosine ethyl, unii-17rpw76a9g, s-ethyl 2-amino-3-4-hydroxyphenyl propanoate | |
| CHEBI:9798 | |
| CCOC(=O)[C@@H](N)CC1=CC=C(O)C=C1 |
Specifications
| 949-67-7 | |
| White-Yellow | |
| MFCD00063046 | |
| l-tyrosine ethyl ester, ethyl l-tyrosinate, tyrosine ethyl ester, h-tyr-oet, l-tyrosine, ethyl ester, tyr-oet, ethyl tyrosine ester, l-tyrosine ethyl, unii-17rpw76a9g, s-ethyl 2-amino-3-4-hydroxyphenyl propanoate | |
| CCOC(=O)[C@@H](N)CC1=CC=C(O)C=C1 | |
| 209.25 | |
| CHEBI:9798 | |
| ≥98.0% (T) | |
| L-Tyrosine Ethyl Ester |
| 103°C | |
| C11H15NO3 | |
| 1 g | |
| SBBWEQLNKVHYCX-JTQLQIEISA-N | |
| ethyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate | |
| 70364 | |
| 209.25 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-1580
TSCA : Yes