missing translation for 'onlineSavingsMsg'
Learn More

L-Tyrosine Ethyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. T098225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T098225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T098225G Supplier TCI America Supplier No. T098225G

Chemical Identifiers

CAS 4089-07-0
Molecular Formula C11H16ClNO3
Molecular Weight (g/mol) 245.70
MDL Number MFCD00063047
InChI Key BQULAXAVRFIAHN-OXZMBGLCNA-N
Synonym l-tyrosine ethyl ester hydrochloride, h-tyr-oet.hcl, ethyl l-tyrosinate hydrochloride, unii-fap58daz1e, s-ethyl 2-amino-3-4-hydroxyphenyl propanoate hydrochloride, h-tyr-oet hcl, tyrosine ethyl ester hydrochloride, fap58daz1e, tyrosine, ethyl ester, hydrochloride, l-tyrosine, ethyl ester hydrochloride
PubChem CID 2724939
IUPAC Name ethyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate hydrochloride
SMILES Cl.CCOC(=O)[C@@H](N)CC1=CC=C(O)C=C1

Specifications

Melting Point 166°C
Color White
Quantity 25 g
Formula Weight 245.70
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material L-Tyrosine Ethyl Ester Hydrochloride
RTECSNumber YP2580000
TSCA No
Recommended Storage Refrigerator