missing translation for 'onlineSavingsMsg'
Learn More

L-Tyrosine ethyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Catalog No. p-7024129
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1835822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1835822 Supplier Thermo Scientific Chemicals Supplier No. A1835822

It is used as a medical and organic intermediate and as an important amino protective agent. It is also used to introduce t-Boc protect gene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a medical and organic intermediate and as an important amino protective agent. It is also used to introduce t-Boc protect gene.

Solubility
Soluble in water (3.685e+005 mg/L @ 25°C (est.)).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 4089-07-0
Molecular Formula C11H16ClNO3
Molecular Weight (g/mol) 245.70
MDL Number MFCD00063047
InChI Key BQULAXAVRFIAHN-OXZMBGLCNA-N
Synonym l-tyrosine ethyl ester hydrochloride, h-tyr-oet.hcl, ethyl l-tyrosinate hydrochloride, unii-fap58daz1e, s-ethyl 2-amino-3-4-hydroxyphenyl propanoate hydrochloride, h-tyr-oet hcl, tyrosine ethyl ester hydrochloride, fap58daz1e, tyrosine, ethyl ester, hydrochloride, l-tyrosine, ethyl ester hydrochloride
PubChem CID 2724939
IUPAC Name ethyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate;hydrochloride
SMILES Cl.CCOC(=O)[C@@H](N)CC1=CC=C(O)C=C1

Specifications

Melting Point 166°C to 168°C
Beilstein 4725904
Solubility Information Soluble in water (3.685e+005mg/L @ 25°C (est.)).
Quantity 100 g
Formula Weight 245.71
Optical Rotation −6.8° (c=2 in Water)
Percent Purity 99%
Chemical Name or Material L-Tyrosine ethyl ester hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 223-820-2
RTECSNumber YP2580000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only