missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Tyrosine Disodium Salt, MP Biomedicals
Specifications
Specifications
| Quantity | 100 g |
| Formula Weight | 225.2 |
| Chemical Name or Material | L-Tyrosine Disodium Salt |
Safety and Handling
| Recommended Storage | 0 to 5°C |