missing translation for 'onlineSavingsMsg'
Learn More

L-Tyrosine Disodium Salt Hydrate 98.0+%, TCI America™

Catalog No. T142325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T142325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T142325G Supplier TCI America Supplier No. T142325G

Chemical Identifiers

CAS 69847-45-6
Molecular Formula C9H9NNa2O3
Molecular Weight (g/mol) 225.16
MDL Number MFCD00037203
InChI Key ASIYFCYUCMQNGK-RIYJSUBBNA-L
Synonym l-tyrosine disodium salt hydrate, l-3-4-hydroxyphenyl alanine disodium salt hydrate, c9h9no3.2na.h2o, l-tyrosine disodium salt hydrate hplc , powder, l-tyrosine disodium salt hydrate, bioreagent, suitable for cell culture, suitable for insect cell culture hplc
PubChem CID 66602062
IUPAC Name disodium (2S)-2-amino-3-(4-oxidophenyl)propanoate
SMILES [Na+].[Na+].N[C@@H](CC1=CC=C([O-])C=C1)C([O-])=O

Specifications

Color Yellow
Quantity 25 g
Formula Weight 225.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material L-Tyrosine Disodium Salt Hydrate
TSCA No
Recommended Storage Refrigerator