Learn More
Thermo Scientific Chemicals L-Tyrosine disodium salt dihydrate
CAS: 122666-87-9 | C9H13NNa2O5 | 261.19 g/mol
$212.40 - $896.59
Chemical Identifiers
| CAS | 122666-87-9 |
|---|---|
| Molecular Formula | C9H13NNa2O5 |
| Molecular Weight (g/mol) | 261.19 |
| MDL Number | MFCD03840558 |
| InChI Key | WFTCFVUQORELJZ-CMIWSEPMNA-L |
| Synonym | tyrosine disodium dihydrate, unii-5rfd27dq22, l-tyrosine disodium salt dihydrate, l-tyrosine, sodium salt, hydrate 1:2:2, c9h9no3.2na.2h2o, disodium l-tyrosinate dihydrate |
| PubChem CID | 71587456 |
| SMILES | O.O.[Na+].[Na+].N[C@@H](CC1=CC=C([O-])C=C1)C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|
AAJ6177022
|
Thermo Scientific Chemicals
J6177022 |
100 g |
Each for $212.40
|
|
|||||
|
AAJ6177036
|
Thermo Scientific Chemicals
J6177036 |
500 g |
Each for $520.55
|
|
|||||
|
AAJ61770A1
|
Thermo Scientific Chemicals
J61770A1 |
1 kg |
Each for $896.59
|
|
|||||
Description
Tyrosine is a polar, naturally occurring, non-essential amino acid. It is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTyrosine is a polar, naturally occurring, non-essential amino acid. It is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.
Solubility
Soluble in water.
Notes
Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
| 122666-87-9 | |
| 261.19 | |
| WFTCFVUQORELJZ-CMIWSEPMNA-L | |
| 71587456 |
| C9H13NNa2O5 | |
| MFCD03840558 | |
| tyrosine disodium dihydrate, unii-5rfd27dq22, l-tyrosine disodium salt dihydrate, l-tyrosine, sodium salt, hydrate 1:2:2, c9h9no3.2na.2h2o, disodium l-tyrosinate dihydrate | |
| O.O.[Na+].[Na+].N[C@@H](CC1=CC=C([O-])C=C1)C([O-])=O |
Specifications
| 122666-87-9 | |
| C9H13NNa2O5 | |
| tyrosine disodium dihydrate, unii-5rfd27dq22, l-tyrosine disodium salt dihydrate, l-tyrosine, sodium salt, hydrate 1:2:2, c9h9no3.2na.2h2o, disodium l-tyrosinate dihydrate | |
| WFTCFVUQORELJZ-CMIWSEPMNA-L | |
| 2-amino-3-(4-hydroxyphenyl)propanoic acid dihydrate disodium | |
| 261.19 | |
| −13° (c=4 in Water) | |
| Very Hygroscopic | |
| Powder |
| Brown to White | |
| MFCD03840558 | |
| Soluble in water. | |
| O.O.[Na+].[Na+].N[C@@H](CC1=CC=C([O-])C=C1)C([O-])=O | |
| 100 g | |
| 71587456 | |
| 261.18 (225.15 Anhydrous) | |
| 99% | |
| L-Tyrosine disodium salt dihydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only