missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L-Tyrosine disodium salt dihydrate
Description
Tyrosine is a polar, naturally occurring, non-essential amino acid. It is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tyrosine is a polar, naturally occurring, non-essential amino acid. It is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.
Solubility
Soluble in water.
Notes
Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Tyrosine is a polar, naturally occurring, non-essential amino acid. It is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.
Solubility
Soluble in water.
Notes
Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Color | Brown to White |
| Solubility Information | Soluble in water. |
| IUPAC Name | 2-amino-3-(4-hydroxyphenyl)propanoic acid dihydrate disodium |
| Quantity | 1 kg |
| Formula Weight | 261.18 (225.15 Anhydrous) |
| Optical Rotation | −13° (c=4 in Water) |
| Sensitivity | Very Hygroscopic |
| Physical Form | Powder |
| Chemical Name or Material | L-Tyrosine disodium salt dihydrate |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only